Gene Transcripts
- Gene Symbol
- Nlrp5
- Entrez Gene ID
- 23968
- Gene Name
- NLR family, pyrin domain containing 5
- Gene Aliases
- Mater, Nalp5, Op1, PAN11
- Chromosome Location
- Chr. 7: 23385897 - 23441921 on Build GRCm38
- UniGene ID
- Mm.333653
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY329486.1 |
- |
11, 12 |
2707 |
|
AY329491.1 |
- |
10, 11 |
2557 |
|
- Gene Symbol
- Nlrp5-ps
- Entrez Gene ID
- 100417675
- Gene Name
- NLR family, pyrin domain containing 5, pseudogene
- Gene Aliases
- Gm18756
- Chromosome Location
- Chr. 7: 14530654 - 14623066 on Build GRCm38
- UniGene ID
- Mm.372763
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016782.1 |
- |
8, 9 |
985 |
|
AK140342.1 |
- |
8, 9 |
1232 |
|
BC150849.1 |
- |
6, 7 |
685 |
|
View Full Product Details