Gene Transcripts
- Gene Symbol
- Trav6-3
- Entrez Gene ID
- 328483
- Gene Name
- T cell receptor alpha variable 6-3
- Gene Aliases
- Gm13948, Gm193, Gm4, OTTMUSG00000015160, TCR
- Chromosome Location
- Chr. 14: 53428761 - 53430377 on Build GRCm38
- UniGene ID
- Mm.390680
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK134090.1 |
- |
|
163 |
|
M61133.1 |
- |
2 |
121 |
|
View Full Product Details