Gene Transcripts

Gene Symbol
Vmn2r8
Entrez Gene ID
627479
Gene Name
vomeronasal 2, receptor 8
Gene Aliases
EG627479
Chromosome Location
Chr. 5: 108797193 - 108808754 on Build GRCm38
UniGene ID
Mm.469886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104620.1 NP_001098090.1 2 417

View Full Product Details