Gene Transcripts

Gene Symbol
RPS6KB2
Entrez Gene ID
6199
Gene Name
ribosomal protein S6 kinase B2
Gene Aliases
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
Chromosome Location
Chr. 11: 67428460 - 67435408 on Build GRCh38
UniGene ID
Hs.534345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003952.2 NP_003943.2 8 710
XM_005274164.1 XP_005274221.1 7 658
XM_005274165.4 XP_005274222.1 7 535
XM_006718655.3 XP_006718718.1 9 831
XM_006718656.3 XP_006718719.1 8 779
XM_006718657.1 XP_006718720.1 8 764
XM_017018108.1 XP_016873597.1 9 842
GenBank mRNA AB016869.1 - 8 682
AB019245.1 - 8 677
AB208827.1 - 4 3982
AF076931.1 - 8 663
AF099739.1 - 8 667
AK297527.1 - 9 816
BC000094.2 - 8 683 3508140
BC006106.1 - 8 666 2959036
BC136564.1 - 8 684 9020554
BX337895.2 - 8 713

View Full Product Details