Gene Transcripts

Gene Symbol
Trim43c
Entrez Gene ID
666731
Gene Name
tripartite motif-containing 43C
Gene Aliases
EG666731, Trim43
Chromosome Location
Chr. 9: 88839023 - 88848198 on Build GRCm38
UniGene ID
Mm.446951
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177858.1 NP_001171329.1 7 1451

View Full Product Details