Gene Transcripts

Gene Symbol
H2-Ab1
Entrez Gene ID
14961
Gene Name
histocompatibility 2, class II antigen A, beta 1
Gene Aliases
AI845868, Abeta, H-2Ab, H2-Ab, I-Abeta, IAb, Ia-2, Ia2, Rmcs1
Chromosome Location
Chr. 17: 34263227 - 34269418 on Build GRCm38
UniGene ID
Mm.254067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207105.3 NP_996988.2 4 834
GenBank mRNA AF065912.1 - 4 737
AF065913.1 - 4 764
AK077436.1 - 4 822
AK155573.1 - 4 806
AK156899.1 - 4 806
AK170370.1 - 4 816
AK171005.1 - 4 806
AK171211.1 - 4 810
AK171281.1 - 4 806
AK171290.1 - 4 806
AK171298.1 - 4 806
AK172409.1 - 4 807
AY452202.1 - 4 743
BC008168.1 - 4 826 3257494
BC010322.1 - 4 800 2651058
BC057998.1 - 4 792 5340289
K01143.1 - 1 109
M13537.1 - 4 749
M13538.1 - 4 743
M13539.1 - 4 743
M13540.1 - 4 743
M15848.1 - 4 779
M21932.1 - 3 649
M66213.1 - 4 743

View Full Product Details