Gene Transcripts

Gene Symbol
Ccdc85c
Entrez Gene ID
668158
Gene Name
coiled-coil domain containing 85C
Gene Aliases
EG668158, Gm265, Gm9010
Chromosome Location
Chr. 12: 108203599 - 108275671 on Build GRCm38
UniGene ID
Mm.482792
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159910.1 NP_001153382.1 4 1321

View Full Product Details