Gene Transcripts

Gene Symbol
Trav6-3
Entrez Gene ID
328483
Gene Name
T cell receptor alpha variable 6-3
Gene Aliases
Gm13948, Gm193, Gm4, OTTMUSG00000015160, TCR
Chromosome Location
Chr. 14: 53428761 - 53430377 on Build GRCm38
UniGene ID
Mm.390680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK134090.1 - 684

View Full Product Details