Gene Transcripts
- Gene Symbol
- Atp2b2
- Entrez Gene ID
- 24215
- Gene Name
- ATPase, Ca++ transporting, plasma membrane 2
- Gene Aliases
- PMCA2
- Chromosome Location
- Chr. 4: 145704779 - 145948997 on Build Rnor_6.0
- UniGene ID
- Rn.90982
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012508.5 |
NP_036640.1
|
|
3262 |
|
GenBank mRNA |
J03754.1 |
- |
|
3268 |
|
View Full Product Details