Gene Transcripts

Gene Symbol
Hprt1
Entrez Gene ID
24465
Gene Name
hypoxanthine phosphoribosyltransferase 1
Gene Aliases
Hgprtase, Hprt
Chromosome Location
Chr. X: 158196640 - 158228815 on Build Rnor_6.0
UniGene ID
Rn.47
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012583.2 NP_036715.1 8 643
GenBank mRNA BC098629.1 - 8 643
FQ212770.1 - 227
FQ212939.1 - 227
FQ213155.1 - 227
FQ215291.1 - 227
FQ215779.1 - 227
FQ216318.1 - 227
FQ219798.1 - 227
FQ219974.1 - 227
FQ220004.1 - 227
FQ229708.1 - 227
FQ230307.1 - 227
FQ234297.1 - 227
M63983.1 - 8 675
M86443.1 - 403
S79292.1 - 8 675
X62085.1 - 603

Gene Symbol
LOC103689983
Entrez Gene ID
103689983
Gene Name
hypoxanthine-guanine phosphoribosyltransferase
Gene Aliases
Chromosome Location
Chr. X: 158623231 - 158655274 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773659.1 XP_008771881.1 8 719

View Full Product Details