Gene Transcripts

Gene Symbol
Gnal
Entrez Gene ID
24611
Gene Name
guanine nucleotide binding protein, alpha stimulating, olfactory type
Gene Aliases
Golf, Olf, RGD1305940
Chromosome Location
Chr. 18: 62805410 - 62944630 on Build Rnor_6.0
UniGene ID
Rn.203753
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191836.1 NP_001178765.1 8, 9 1048
GenBank mRNA S80376.1 - 8, 9 1450

View Full Product Details