Gene Transcripts

Gene Symbol
Pld1
Entrez Gene ID
25096
Gene Name
phospholipase D1
Gene Aliases
Pld1a, Pld1b, Plda, Pldb
Chromosome Location
Chr. 2: 113651964 - 113849403 on Build Rnor_6.0
UniGene ID
Rn.11130
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030992.1 NP_112254.1 14 1586
XM_006232205.2 XP_006232267.1 14 2361
XM_006232206.2 XP_006232268.1 15 2014
XM_008760891.1 XP_008759113.1 15 2014
GenBank mRNA AB000778.1 - 14 1601
AB000779.1 - 14 1586
AB003170.1 - 14 1478
AB003171.1 - 14 1478
AF017251.1 - 13 1430
U69550.1 - 14 1586
U88986.1 - 14 1498

View Full Product Details