Gene Transcripts

Gene Symbol
Inhbb
Entrez Gene ID
25196
Gene Name
inhibin beta-B
Gene Aliases
Chromosome Location
Chr. 13: 35436532 - 35442222 on Build Rnor_6.0
UniGene ID
Rn.35074
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080771.1 NP_542949.1 2 516
GenBank mRNA AF478684.1 - 1 47

View Full Product Details