Gene Transcripts
- Gene Symbol
- Bsg
- Entrez Gene ID
- 25246
- Gene Name
- basigin (Ok blood group)
- Gene Aliases
- 5A11, CE9, EMMPRIN, HT7, Ox47R, basignin
- Chromosome Location
- Chr. 7: 12875536 - 12882753 on Build Rnor_6.0
- UniGene ID
- Rn.2269
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001109882.1 |
NP_001103352.1
|
3, 4 |
665 |
|
NM_012783.3 |
NP_036915.1
|
2, 3 |
317 |
|
GenBank mRNA |
AY120888.1 |
- |
3, 4 |
584 |
|
BC059145.1 |
- |
2, 3 |
241 |
6889785 |
CV119278.1 |
- |
3, 4 |
665 |
|
FQ212571.1 |
- |
2, 3 |
253 |
|
FQ212582.1 |
- |
2, 3 |
261 |
|
FQ213065.1 |
- |
2, 3 |
263 |
|
FQ213626.1 |
- |
2, 3 |
253 |
|
FQ214193.1 |
- |
2, 3 |
253 |
|
FQ214232.1 |
- |
2, 3 |
258 |
|
FQ218399.1 |
- |
2, 3 |
253 |
|
FQ219910.1 |
- |
2, 3 |
237 |
|
FQ220920.1 |
- |
2, 3 |
261 |
|
FQ224837.1 |
- |
2, 3 |
262 |
|
FQ229144.1 |
- |
2, 3 |
263 |
|
FQ229755.1 |
- |
2, 3 |
250 |
|
FQ230014.1 |
- |
2, 3 |
263 |
|
FQ231850.1 |
- |
2, 3 |
263 |
|
FQ232262.1 |
- |
2, 3 |
253 |
|
FQ234420.1 |
- |
2, 3 |
263 |
|
FQ234809.1 |
- |
2, 3 |
261 |
|
X54640.1 |
- |
|
444 |
|
X67215.1 |
- |
2, 3 |
238 |
|
View Full Product Details