Gene Transcripts

Gene Symbol
Bsg
Entrez Gene ID
25246
Gene Name
basigin (Ok blood group)
Gene Aliases
5A11, CE9, EMMPRIN, HT7, Ox47R, basignin
Chromosome Location
Chr. 7: 12875536 - 12882753 on Build Rnor_6.0
UniGene ID
Rn.2269
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109882.1 NP_001103352.1 6 968
NM_012783.3 NP_036915.1 5 620
GenBank mRNA AY120888.1 - 6 887
BC059145.1 - 5 544 6889785
FQ212571.1 - 5 556
FQ212582.1 - 5 564
FQ213065.1 - 5 566
FQ213626.1 - 5 556
FQ214193.1 - 5 556
FQ214232.1 - 5 561
FQ219910.1 - 5 540
FQ219990.1 - 5 556
FQ220920.1 - 5 564
FQ224837.1 - 5 565
FQ229144.1 - 5 566
FQ229755.1 - 5 553
FQ230014.1 - 5 566
FQ231850.1 - 5 566
FQ232262.1 - 5 556
FQ234420.1 - 5 566
FQ234809.1 - 5 564
FQ234829.1 - 5 556
X54640.1 - 747
X67215.1 - 5 541

View Full Product Details