Gene Transcripts

Gene Symbol
Cyp2c6v1
Entrez Gene ID
293989
Gene Name
cytochrome P450, family 2, subfamily C, polypeptide 6, variant 1
Gene Aliases
CYPIIC6, Cyp2c6, PB1, PTF2
Chromosome Location
Chr. 1: 147157097 - 147158284 on Build Rnor_6.0
UniGene ID
Rn.91122
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759613.1 XP_008757835.1 5, 6 863
XM_008759614.1 XP_008757836.1 6, 7 863
XM_008759615.1 XP_008757837.1 5, 6 863

View Full Product Details