Gene Transcripts

Gene Symbol
Nme6
Entrez Gene ID
58964
Gene Name
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases
Nm23-R6
Chromosome Location
Chr. 8: 117920912 - 117928148 on Build Rnor_6.0
UniGene ID
Rn.36170
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191884.1 NP_001178813.1 7 646
XM_006243847.2 XP_006243909.1 7 655
XM_006243848.2 XP_006243910.1 7 581
XM_006243849.1 XP_006243911.1 7 1099
XM_006243850.2 XP_006243912.1 7 620
XM_006243851.1 XP_006243913.1 7 684
GenBank mRNA AF051943.1 - 5 388
FQ225386.1 - 7 825

View Full Product Details