Gene Transcripts

Gene Symbol
Sik1
Entrez Gene ID
59329
Gene Name
salt-inducible kinase 1
Gene Aliases
Sik, Snf1lk
Chromosome Location
Chr. 20: 10670747 - 10680279 on Build Rnor_6.0
UniGene ID
Rn.42905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021693.2 NP_067725.2 12 1810
GenBank mRNA AB020480.1 - 12 1810
AF106937.1 - 1868

View Full Product Details