Gene Transcripts

Gene Symbol
HSPA13
Entrez Gene ID
6782
Gene Name
heat shock protein family A (Hsp70) member 13
Gene Aliases
STCH
Chromosome Location
Chr. 21: 14371115 - 14383188 on Build GRCh38
UniGene ID
Hs.352341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006948.4 NP_008879.3 3 525
GenBank mRNA AK223282.1 - 3 483
AK312396.1 - 3 480
BC036370.2 - 3 487 4837136
U04735.1 - 3 492

View Full Product Details