Gene Transcripts

Gene Symbol
Ihh
Entrez Gene ID
84399
Gene Name
Indian hedgehog
Gene Aliases
Chromosome Location
Chr. 9: 82208223 - 82214440 on Build Rnor_6.0
UniGene ID
Rn.8711
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053384.1 NP_445836.1 2 712
GenBank mRNA AF162914.1 - 1 181

View Full Product Details