Gene Transcripts

Gene Symbol
Rab9a
Entrez Gene ID
84589
Gene Name
RAB9A, member RAS oncogene family
Gene Aliases
Rab9
Chromosome Location
Chr. X: 29525256 - 29547295 on Build Rnor_6.0
UniGene ID
Rn.35289
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053458.1 NP_445910.1 3 633
GenBank mRNA AF325692.1 - 3 633
BC070502.1 - 3 637 7096737
FQ212962.1 - 3 642

View Full Product Details