Gene Transcripts

Gene Symbol
Il2rg
Entrez Gene ID
140924
Gene Name
interleukin 2 receptor, gamma
Gene Aliases
Ab2-183, Cd132
Chromosome Location
Chr. X: 71165378 - 71169038 on Build Rnor_6.0
UniGene ID
Rn.14508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080889.1 NP_543165.1 2 157
GenBank mRNA AF410926.1 - 2 77
BC079343.1 - 2 157 7106866

View Full Product Details