Gene Transcripts
- Gene Symbol
- Obp1f
- Entrez Gene ID
- 192267
- Gene Name
- odorant binding protein I f
- Gene Aliases
- Obp-1f
- Chromosome Location
- Chr. X: 45271742 - 45284357 on Build Rnor_6.0
- UniGene ID
- Rn.9718
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100362409
- Entrez Gene ID
- 100362409
- Gene Name
- odorant-binding protein-like
- Gene Aliases
- Chromosome Location
- Chr. X: 83859898 - 83864401 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC103690319
- Entrez Gene ID
- 103690319
- Gene Name
- odorant-binding protein
- Gene Aliases
- Chromosome Location
- Chr. X: 45212961 - 45218173 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details