Gene Transcripts

Gene Symbol
Akr1b8
Entrez Gene ID
286921
Gene Name
aldo-keto reductase family 1, member B8
Gene Aliases
Chromosome Location
Chr. 4: 61772064 - 61784637 on Build Rnor_6.0
UniGene ID
Rn.23676
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173136.1 NP_775159.1 8 814
GenBank mRNA AJ277957.1 - 8 814

View Full Product Details