Gene Transcripts

Gene Symbol
Slc16a11
Entrez Gene ID
287450
Gene Name
solute carrier family 16, member 11
Gene Aliases
Chromosome Location
Chr. 10: 56822756 - 56826480 on Build Rnor_6.0
UniGene ID
Rn.89414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105797.2 NP_001099267.2 6 1095
XM_006246646.1 XP_006246708.1 6 1235
XM_006246647.2 XP_006246709.1 5 1778
XM_006246648.2 XP_006246710.1 5 1080
XM_006246649.2 XP_006246711.1 4 799
XM_008767805.1 XP_008766027.1 5 1639
GenBank mRNA BC169003.1 - 6 1235
FQ219143.1 - 3 570

View Full Product Details