Gene Transcripts

Gene Symbol
Wbscr28
Entrez Gene ID
288604
Gene Name
Williams-Beuren syndrome chromosome region 28
Gene Aliases
RGD1561903
Chromosome Location
Chr. 12: 24802646 - 24808287 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249213.2 XP_006249275.1 3 1412
XM_213758.8 XP_213758.4 3 771

View Full Product Details