Gene Transcripts

Gene Symbol
Tesc
Entrez Gene ID
288689
Gene Name
tescalcin
Gene Aliases
RGD1566317
Chromosome Location
Chr. 12: 44141258 - 44174701 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249426.2 XP_006249488.1 4 368
XM_213790.9 XP_213790.6 3 269

View Full Product Details