Gene Transcripts

Gene Symbol
Gadd45gip1
Entrez Gene ID
288916
Gene Name
growth arrest and DNA-damage-inducible, gamma interacting protein 1
Gene Aliases
MRP-L59
Chromosome Location
Chr. 19: 25946979 - 25949528 on Build Rnor_6.0
UniGene ID
Rn.161715
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100504.1 NP_001093974.1 2 436
GenBank mRNA BC083178.1 - 2 428

View Full Product Details