Gene Transcripts

Gene Symbol
TTF1
Entrez Gene ID
7270
Gene Name
transcription termination factor 1
Gene Aliases
TTF-1, TTF-I
Chromosome Location
Chr. 9: 132375548 - 132406851 on Build GRCh38
UniGene ID
Hs.54780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205296.1 NP_001192225.1 5 609
NM_007344.3 NP_031370.2 6 1983
NR_134525.1 6 1983
XM_006717273.3 XP_006717336.2 6 1991
XM_017015105.1 XP_016870594.1 6 2490
GenBank mRNA AK302902.1 - 6 1969
BC127669.1 - 5 584
BC127670.1 - 5 584
BC143048.1 - 5 584
BC143049.1 - 5 584
BC143821.1 - 6 1956

View Full Product Details