Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003320.4 | NP_003311.2 | 9, 10 | 1415 | |
NM_177972.2 | NP_813977.1 | 8, 9 | 1158 | ||
XM_005253109.3 | XP_005253166.1 | 8, 9 | 1150 | ||
XM_011520344.2 | XP_011518646.1 | 8, 9 | 1205 | ||
GenBank mRNA | AB209618.1 | - | 1 | 5832 | |
AK293528.1 | - | 8, 9 | 1086 | ||
BC075031.2 | - | 9, 10 | 1207 | 30915625 | |
BC075032.2 | - | 9, 10 | 1207 | 30915418 | |
U54644.1 | - | 8, 9 | 1161 | ||
U82467.1 | - | 9, 10 | 1415 |