Gene Transcripts

Gene Symbol
Aga
Entrez Gene ID
290923
Gene Name
aspartylglucosaminidase
Gene Aliases
Chromosome Location
Chr. 16: 41222225 - 41234163 on Build Rnor_6.0
UniGene ID
Rn.104649
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031641.1 NP_001026811.1 6, 7 699
XM_006253111.2 XP_006253173.1 7, 8 824
GenBank mRNA BC098718.1 - 6, 7 699

View Full Product Details