Gene Transcripts

Gene Symbol
Dsg1
Entrez Gene ID
291755
Gene Name
desmoglein 1
Gene Aliases
Dsg1b, Dsg1c
Chromosome Location
Chr. 18: 11970118 - 12040337 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771993.1 XP_008770215.1 18 3032

View Full Product Details