Gene Transcripts

Gene Symbol
Sult5a1
Entrez Gene ID
292077
Gene Name
sulfotransferase family 5A, member 1
Gene Aliases
Chromosome Location
Chr. 19: 55952861 - 55978583 on Build Rnor_6.0
UniGene ID
Rn.107747
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106194.1 NP_001099664.1 3 317
NM_001201369.1 NP_001188298.1 4 466
GenBank mRNA FM102597.1 - 4 466

View Full Product Details