Gene Transcripts

Gene Symbol
Fpr-rs3
Entrez Gene ID
292419
Gene Name
formyl peptide receptor, related sequence 3
Gene Aliases
Chromosome Location
Chr. 1: 59902308 - 59903339 on Build Rnor_6.0
UniGene ID
Rn.222430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169140.1 NP_001162611.1 1 576

View Full Product Details