Gene Transcripts

Gene Symbol
Vom2r28
Entrez Gene ID
292547
Gene Name
vomeronasal 2 receptor, 28
Gene Aliases
RGD1565786
Chromosome Location
Chr. 1: 69558008 - 69589096 on Build Rnor_6.0
UniGene ID
Rn.217628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099472.1 NP_001092942.1 3 351

View Full Product Details