Gene Transcripts

Gene Symbol
Qpctl
Entrez Gene ID
292687
Gene Name
glutaminyl-peptide cyclotransferase-like
Gene Aliases
RGD1308128
Chromosome Location
Chr. 1: 80047504 - 80057016 on Build Rnor_6.0
UniGene ID
Rn.100458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106230.1 NP_001099700.1 4, 5 843
XM_006228402.2 XP_006228464.1 4, 5 1285
GenBank mRNA BC169076.1 - 4, 5 839

View Full Product Details