Gene Transcripts

Gene Symbol
Klk13
Entrez Gene ID
292848
Gene Name
kallikrein related-peptidase 13
Gene Aliases
Chromosome Location
Chr. 1: 99648658 - 99660291 on Build Rnor_6.0
UniGene ID
Rn.222426
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170405.1 NP_001163876.1 3 265
XM_008759331.1 XP_008757553.1 2 184

View Full Product Details