Gene Transcripts

Gene Symbol
Tbc1d17
Entrez Gene ID
292886
Gene Name
TBC1 domain family, member 17
Gene Aliases
Chromosome Location
Chr. 1: 100836993 - 100845216 on Build Rnor_6.0
UniGene ID
Rn.203954
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106258.2 NP_001099728.1 11 1182
GenBank mRNA BC160880.1 - 11 1182

View Full Product Details