Gene Transcripts

Gene Symbol
Olr92
Entrez Gene ID
293233
Gene Name
olfactory receptor 92
Gene Aliases
Chromosome Location
Chr. 1: 168455308 - 168456252 on Build Rnor_6.0
UniGene ID
Rn.141895
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000140.1 NP_001000140.1 1 208

View Full Product Details