Gene Transcripts

Gene Symbol
Bcl7c
Entrez Gene ID
293514
Gene Name
B-cell CLL/lymphoma 7C
Gene Aliases
Chromosome Location
Chr. 1: 199155296 - 199159125 on Build Rnor_6.0
UniGene ID
Rn.92046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106298.1 NP_001099768.1 4 581
XM_006230256.2 XP_006230318.1 4 477

View Full Product Details