Gene Transcripts

Gene Symbol
Dnah3
Entrez Gene ID
117249
Gene Name
dynein, axonemal, heavy chain 3
Gene Aliases
Dnahc3, Dnahc3l
Chromosome Location
Chr. 1: 189688826 - 189865233 on Build Rnor_6.0
UniGene ID
Rn.92386
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759836.1 XP_008758058.1 51 8549

View Full Product Details