Gene Transcripts

Gene Symbol
Col17a1
Entrez Gene ID
294027
Gene Name
collagen, type XVII, alpha 1
Gene Aliases
Chromosome Location
Chr. 1: 267416681 - 267464451 on Build Rnor_6.0
UniGene ID
Rn.40392
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106366.1 NP_001099836.1 48 3894
XM_006231476.2 XP_006231538.1 49 4754

View Full Product Details