Gene Transcripts

Gene Symbol
Aire
Entrez Gene ID
294328
Gene Name
autoimmune regulator
Gene Aliases
Chromosome Location
Chr. 20: 11365630 - 11380636 on Build Rnor_6.0
UniGene ID
Rn.218492
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106379.1 NP_001099849.1 2 302
XM_006256247.2 XP_006256309.1 2 369

View Full Product Details