Gene Transcripts

Gene Symbol
Aadacl2
Entrez Gene ID
295076
Gene Name
arylacetamide deacetylase-like 2
Gene Aliases
RGD1563197
Chromosome Location
Chr. 2: 150344307 - 150365033 on Build Rnor_6.0
UniGene ID
Rn.228196
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191706.1 NP_001178635.1 543

Gene Symbol
LOC100910567
Entrez Gene ID
100910567
Gene Name
arylacetamide deacetylase-like 2-like
Gene Aliases
Chromosome Location
Chr. 2: 150106180 - 150127100 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749284.2 XP_003749332.1 4 655

View Full Product Details