Gene Transcripts

Gene Symbol
Apoa1bp
Entrez Gene ID
295229
Gene Name
apolipoprotein A-I binding protein
Gene Aliases
AI-BP
Chromosome Location
Chr. 2: 187424319 - 187426410 on Build Rnor_6.0
UniGene ID
Rn.14886
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106440.1 NP_001099910.1 1 308
XM_006232609.2 XP_006232671.1 2 250
GenBank mRNA BC158876.1 - 2 215

View Full Product Details