Gene Transcripts

Gene Symbol
Tnks1bp1
Entrez Gene ID
295707
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
Chromosome Location
Chr. 3: 72458539 - 72483206 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006234484.2 XP_006234546.1 3 714
XM_006234485.2 XP_006234547.1 3 1194
XM_006234486.2 XP_006234548.1 3 657

View Full Product Details