Gene Transcripts

Gene Symbol
Bpifb2
Entrez Gene ID
296290
Gene Name
BPI fold containing family B, member 2
Gene Aliases
Bpil1, Lplunc2
Chromosome Location
Chr. 3: 149358881 - 149377867 on Build Rnor_6.0
UniGene ID
Rn.162713
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106531.1 NP_001100001.1 9, 10 1312

View Full Product Details