Gene Transcripts

Gene Symbol
Dolpp1
Entrez Gene ID
296624
Gene Name
dolichyldiphosphatase 1
Gene Aliases
Chromosome Location
Chr. 3: 8957096 - 8966618 on Build Rnor_6.0
UniGene ID
Rn.93101
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106567.1 NP_001100037.1 1 256
XM_006233783.2 XP_006233845.1 1 1293

View Full Product Details