Gene Transcripts

Gene Symbol
ZNF32
Entrez Gene ID
7580
Gene Name
zinc finger protein 32
Gene Aliases
KOX30
Chromosome Location
Chr. 10: 43643859 - 43649350 on Build GRCh38
UniGene ID
Hs.522885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005368.2 NP_001005368.1 3 551
NM_001324164.1 NP_001311093.1 3 519
NM_001324165.1 NP_001311094.1 3 657
NM_001324166.1 NP_001311095.1 3 421
NM_001324167.1 NP_001311096.1 3 417
NM_001324168.1 NP_001311097.1 3 555
NM_006973.2 NP_008904.1 3 413
XM_017016616.1 XP_016872105.1 2 3484
GenBank mRNA AI871265.1 - 3 456
AK312132.1 - 3 410
BC022842.1 - 2 348 5014207
BG477527.1 - 3 415
U69645.1 - 3 389
X52361.1 - 1 39

View Full Product Details