Gene Transcripts

Gene Symbol
Magoh
Entrez Gene ID
298385
Gene Name
mago-nashi homolog, proliferation-associated (Drosophila)
Gene Aliases
Chromosome Location
Chr. 5: 127480468 - 127487617 on Build Rnor_6.0
UniGene ID
Rn.8332
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100536.1 NP_001094006.1 3, 4 333
XM_006238510.2 XP_006238572.1 3, 4 425
GenBank mRNA DQ359101.1 - 3, 4 262

View Full Product Details